missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasminogen Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB300-544-0.025mg
This item is not returnable.
View return policy
Description
Plasminogen Polyclonal specifically detects Plasminogen in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Plasminogen | |
| Polyclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
| DKFZp779M0222, EC 3.4.21, EC 3.4.21.7, plasmin, plasminogen | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse and Human.Predicted cross-reactivity based on antigen sequence identity: Rat (95%). | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2 μg/mL, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin | |
| P20918 | |
| PLG | |
| Synthetic Peptide: V(98) Y L S E C K T G I G N G Y R G T M S(116) | |
| 0.025 mg | |
| Cell Cycle and Replication | |
| 5340 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction