missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasminogen Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
200.00€
Specifications
| Antigen | Plasminogen |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 μg/mL, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Plasminogen Polyclonal specifically detects Plasminogen in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Plasminogen | |
| Western Blot 2 μg/mL, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| P20918 | |
| 5340 | |
| Synthetic Peptide: V(98) Y L S E C K T G I G N G Y R G T M S(116) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
| DKFZp779M0222, EC 3.4.21, EC 3.4.21.7, plasmin, plasminogen | |
| PLG | |
| IgG | |
| Immunogen affinity purified | |
| Mouse and Human.Predicted cross-reactivity based on antigen sequence identity: Rat (95%). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title