missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pit1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
201.00€ - 441.00€
Specifications
| Antigen | Pit1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30226357
|
Novus Biologicals
NBP3-38647SS |
0.025 mg |
201.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30226356
|
Novus Biologicals
NBP3-38647 |
0.1 mg |
441.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pit1 Polyclonal antibody specifically detects Pit1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)Specifications
| Pit1 | |
| Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| P28069 | |
| 5449 | |
| IgG | |
| Immunogen affinity purified |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cell Cycle and Replication | |
| PBS | |
| CPHD1, GHF1, GHF-1pituitary-specific positive transcription factor 1, Growth hormone factor 1, PIT-1, PIT1pituitary-specific transcription factor 1, POU class 1 homeobox 1, POU domain class 1, transcription factor 1, POU domain, class 1, transcription factor 1, POU1F1a | |
| Partial recombinant protein made to an internal portion of the human Pit1 protein [UniProt accession number P28069]. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title