missing translation for 'onlineSavingsMsg'
Learn More

Pit1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-38647SS

 View more versions of this product

Product Code. 30226357

  • 201.00€ / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Pit1 Polyclonal antibody specifically detects Pit1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pit1
Polyclonal
Unconjugated
PBS
CPHD1, GHF1, GHF-1pituitary-specific positive transcription factor 1, Growth hormone factor 1, PIT-1, PIT1pituitary-specific transcription factor 1, POU class 1 homeobox 1, POU domain class 1, transcription factor 1, POU domain, class 1, transcription factor 1, POU1F1a
Partial recombinant protein made to an internal portion of the human Pit1 protein [UniProt accession number P28069].
0.025 mg
Breast Cancer, Cell Cycle and Replication
5449
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
1.0 mg/mL
Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200
P28069
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.