missing translation for 'onlineSavingsMsg'
Learn More

PIP2 Antibody (PIP2 2C11), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

Brand:  Novus Biologicals NBP2-76433

 View more versions of this product

Product Code. 18098274

  • 412.00€ / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PIP2 Monoclonal specifically detects PIP2 in Non-species specific samples. It is validated for Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PIP2
Monoclonal
1 mg/ml
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional, Dot Blot, Chromatin Immunoprecipitation (ChIP)
1,2-Diacyl-sn-glycero-3-phospho-(1-D-myo-inositol 4,5-bisphosphate), Phosphatidylinositol 4,5-bisphosphate, Phosphatidylinositol biphosphate, Phosphatidylinositol diphosphate, PIP 2, PtdIns(4,5)P2
Liposomes (prepared in PBS with lipid A, phosphatidylcholine (PC) and cholesterol) containing synthetic di-palmitoyl PtdIns(4,5)P2.
0.1 mg
Cross reacts with PtdIns(4,5)P2, Ptd(4)P and Ptd(3,4,5)P3.
Store at -20°C. Avoid freeze-thaw cycles.
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Dot Blot
PIP2 2C11
Unconjugated
PBS with 0.02% Sodium Azide
Mouse
Thiophilic Chromatography
Primary
Non-species specific
IgM
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.