missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP2 Antibody (PIP2 2C11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
214.00€ - 412.00€
Specifications
| Antigen | PIP2 |
|---|---|
| Clone | PIP2 2C11 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18098274
|
Novus Biologicals
NBP2-76433 |
0.1 mg |
412.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654819
|
Novus Biologicals
NBP2-76433-0.025mg |
0.025 mg |
214.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PIP2 Monoclonal specifically detects PIP2 in Non-species specific samples. It is validated for Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional.Specifications
| PIP2 | |
| Monoclonal | |
| Mouse | |
| 1,2-Diacyl-sn-glycero-3-phospho-(1-D-myo-inositol 4,5-bisphosphate), Phosphatidylinositol 4,5-bisphosphate, Phosphatidylinositol biphosphate, Phosphatidylinositol diphosphate, PIP 2, PtdIns(4,5)P2 | |
| IgM | |
| Thiophilic Chromatography |
| PIP2 2C11 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Liposomes (prepared in PBS with lipid A, phosphatidylcholine (PC) and cholesterol) containing synthetic di-palmitoyl PtdIns(4,5)P2. | |
| Primary | |
| Cross reacts with PtdIns(4,5)P2, Ptd(4)P and Ptd(3,4,5)P3. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title