missing translation for 'onlineSavingsMsg'
Learn More

Piezo1 (extracellular domain) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen Piezo1 (extracellular domain)
Concentration 300 μg/mL
Applications Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16886175
View Documents
Proteintech
28511-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16876175
View Documents
Proteintech
28511-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Piezo1 (extracellular domain) antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Piezo1 (extracellular domain). This antibody recognizes human antigen. The Piezo1 (extracellular domain) antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Specifications

Piezo1 (extracellular domain)
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
Unconjugated
Human
IgG
300 μg/mL
Polyclonal
Rabbit
9780
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.