Learn More
Piezo1 (extracellular domain) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28511-1-AP-20UL
Description
The Piezo1 (extracellular domain) antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Piezo1 (extracellular domain). This antibody recognizes human antigen. The Piezo1 (extracellular domain) antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| Piezo1 (extracellular domain) | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 9780 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human |