missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF13 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
201.00€ - 481.00€
Specifications
| Antigen | PHF13 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693632
|
Novus Biologicals
NBP2-94427-0.02ml |
0.02 mL |
201.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18624692
|
Novus Biologicals
NBP2-94427-0.1ml |
0.1 mL |
481.00€
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PHF13 Polyclonal antibody specifically detects PHF13 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PHF13 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 148479 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| MGC43399, PHD finger protein 13, PHD zinc finger protein PHF5, PHF5, SPOC1, Survival time-associated PHD finger protein in ovarian cancer 1, survival time-associated PHD protein in ovarian cancer | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PHF13 (NP_722519.2). MDSDSCAAAFHPEEYSPSCKRRRTVEDFNKFCTFVLAYAGYIPYPKEELPLRSSPSPANSTAGTIDSDGWDAGFSDIASSVPLPVSDRCFSHLQPTLLQRAKPSNFLLDRKKTDKLKKKKKRKRRDSDAPGKEGYRGGLLKLEAADPYVETPTSPTLQDIPQAPSDPCSGWDSDTPSSGSCATVSPDQVKEIKTEGKRTIVRQGKQVVFRDEDSTGNDEDIMVDSDDDSWDLVTCFCMKPFAGRPMIECNECHTWIHLSCAKIRKSNVPEVFVCQKCRDSKFDIRRSNRSRTGSRKLFLD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title