missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHF13 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94427-0.1ml
This item is not returnable.
View return policy
Description
PHF13 Polyclonal antibody specifically detects PHF13 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PHF13 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC43399, PHD finger protein 13, PHD zinc finger protein PHF5, PHF5, SPOC1, Survival time-associated PHD finger protein in ovarian cancer 1, survival time-associated PHD protein in ovarian cancer | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PHF13 (NP_722519.2). MDSDSCAAAFHPEEYSPSCKRRRTVEDFNKFCTFVLAYAGYIPYPKEELPLRSSPSPANSTAGTIDSDGWDAGFSDIASSVPLPVSDRCFSHLQPTLLQRAKPSNFLLDRKKTDKLKKKKKRKRRDSDAPGKEGYRGGLLKLEAADPYVETPTSPTLQDIPQAPSDPCSGWDSDTPSSGSCATVSPDQVKEIKTEGKRTIVRQGKQVVFRDEDSTGNDEDIMVDSDDDSWDLVTCFCMKPFAGRPMIECNECHTWIHLSCAKIRKSNVPEVFVCQKCRDSKFDIRRSNRSRTGSRKLFLD | |
| 0.1 mL | |
| Cell Biology | |
| 148479 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction