missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAB1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93389-0.1ml
This item is not returnable.
View return policy
Description
NAB1 Polyclonal antibody specifically detects NAB1 in Human samples. It is validated for Western Blot
Specifications
| NAB1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EGR1 binding protein 1, EGR-1-binding protein 1, NGFI-A binding protein 1 (EGR1 binding protein 1), NGFI-A-binding protein 1, Transcriptional regulatory protein p54 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 189-317 of human NAB1 (NP_005957.2). EALDAAAALSVAECVERMAPTLPKSDLNEVKELLKTNKKLAKMIGHIFEMNDDDPHKEEEIRKYSAIYGRFDSKRKDGKHLTLHELTVNEAAAQLCVKDNALLTRRDELFALARQISREVTYKYTYRTT | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 4664 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction