missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAB1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 470.00€
Specifications
| Antigen | NAB1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693580
|
Novus Biologicals
NBP2-93389-0.02ml |
0.02 mL |
190.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623540
|
Novus Biologicals
NBP2-93389-0.1ml |
0.1 mL |
470.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NAB1 Polyclonal antibody specifically detects NAB1 in Human samples. It is validated for Western BlotSpecifications
| NAB1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 4664 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EGR1 binding protein 1, EGR-1-binding protein 1, NGFI-A binding protein 1 (EGR1 binding protein 1), NGFI-A-binding protein 1, Transcriptional regulatory protein p54 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 189-317 of human NAB1 (NP_005957.2). EALDAAAALSVAECVERMAPTLPKSDLNEVKELLKTNKKLAKMIGHIFEMNDDDPHKEEEIRKYSAIYGRFDSKRKDGKHLTLHELTVNEAAAQLCVKDNALLTRRDELFALARQISREVTYKYTYRTT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title