missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX12 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93666-0.1ml
This item is not returnable.
View return policy
Description
PEX12 Polyclonal antibody specifically detects PEX12 in Human samples. It is validated for Western Blot
Specifications
| PEX12 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| PAF3, PAF-3, peroxin 12, Peroxin-12, peroxisomal biogenesis factor 12, Peroxisome assembly factor 3, peroxisome assembly protein 12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 290-359 of human PEX12 (NP_000277.1). YNSDSPLLPKMKTVCPLCRKTRVNDTVLATSGYVFCYRCVFHYVRSHQACPITGYPTEVQHLIKLYSPEN | |
| 0.1 mL | |
| Signal Transduction | |
| 5193 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction