missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00€ - 624.00€
Specifications
| Antigen | PEX10 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18238612
|
Novus Biologicals
NBP2-58492 |
100 μL |
624.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18601509
|
Novus Biologicals
NBP2-58492-25ul |
25 μL |
415.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PEX10 Polyclonal specifically detects PEX10 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| PEX10 | |
| Polyclonal | |
| Rabbit | |
| Zinc Finger | |
| NALD, peroxin 10, Peroxin-10, peroxisomal biogenesis factor 10MGC1998, Peroxisome assembly protein 10, RING finger protein 69, RNF69peroxisome biogenesis factor 10 | |
| PEX10 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 5192 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GITYQALRPDPLRVLMSVAPSALQLRVRSLPGEDLRARVSYR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title