missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periplakin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
201.00€ - 482.00€
Specifications
| Antigen | Periplakin |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100 - 1:500 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631412
|
Novus Biologicals
NBP2-94787-0.02ml |
0.02 mL |
201.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648122
|
Novus Biologicals
NBP2-94787-0.1ml |
0.1 mL |
482.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Periplakin Polyclonal antibody specifically detects Periplakin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Periplakin | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 5493 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 190 kDa paraneoplastic pemphigus antigen, 195 kDa cornified envelope precursor protein, KIAA0568, MGC134872, periplakin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human PPL (NP_002696.3). MNSLFRKRNKGKYSPTVQTRSISNKELSELIEQLQKNADQVEKNIVDTEAKMQSDLARLQEGRQPEHRDVTLQKVLDSEKLLYVLEADAAIAKHMKHPQGDMIAEDIRQLKERVTNLRGKHKQIYRLAVK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title