missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periplakin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94787-0.1ml
408.00 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Periplakin Polyclonal antibody specifically detects Periplakin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Periplakin | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100 - 1:500 | |
| 190 kDa paraneoplastic pemphigus antigen, 195 kDa cornified envelope precursor protein, KIAA0568, MGC134872, periplakin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human PPL (NP_002696.3). MNSLFRKRNKGKYSPTVQTRSISNKELSELIEQLQKNADQVEKNIVDTEAKMQSDLARLQEGRQPEHRDVTLQKVLDSEKLLYVLEADAAIAKHMKHPQGDMIAEDIRQLKERVTNLRGKHKQIYRLAVK | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction | |
| 5493 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction