missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peripherin Antibody (8G2), Novus Biologicals™
Mouse Monoclonal Antibody
222.00€ - 539.00€
Specifications
| Antigen | Peripherin |
|---|---|
| Clone | 8G2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:5000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18040264
|
Novus Biologicals
NB300-138-100UL |
100 μL |
539.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18277882
|
Novus Biologicals
NB300-138-0.025ML |
0.025 mL |
222.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Peripherin Monoclonal specifically detects Peripherin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Peripherin | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Human, Mouse, Rat, Pig, Bovine | |
| Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4 | |
| PRPH | |
| IgG1 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 8G2 | |
| Western Blot 1:5000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500 | |
| Monoclonal | |
| Mouse | |
| PBS, 50% Glycerol with 0.05% sodium azide | |
| 5630 | |
| Recombinant rat Peripherin purified from E. coli. [UniProtNo. P21807] | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title