missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peripherin Antibody (8G2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-138-0.025ML
This item is not returnable.
View return policy
Description
Peripherin Monoclonal specifically detects Peripherin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Peripherin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:5000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence 1:500 | |
| Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4 | |
| Mouse | |
| 0.025 mL | |
| 5630 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| 8G2 | |
| Unconjugated | |
| PBS, 50% Glycerol with 0.05% sodium azide | |
| PRPH | |
| Recombinant rat Peripherin purified from E. coli. [UniProtNo. P21807] | |
| Primary | |
| Rat, Bovine, Mouse, Pig, Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction