missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PELO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | PELO |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16857053
|
Proteintech
10582-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16847053
|
Proteintech
10582-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein which contains a conserved nuclear localization signal. The encoded protein may have a role in spermatogenesis, cell cycle control, and in meiotic cell division.Specifications
| PELO | |
| Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q5XIP1, Q80X73, Q9BRX2 | |
| 105083, 294754, 53918 | |
| PELO Fusion Protein Ag0873 | |
| Primary | |
| -20°C | |
| PELO |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CGI 17, PELO, pelota homolog (Drosophila), PRO1770, Protein pelota homolog | |
| PELO | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title