Learn More
PELO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10582-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein which contains a conserved nuclear localization signal. The encoded protein may have a role in spermatogenesis, cell cycle control, and in meiotic cell division.Specifications
| PELO | |
| Polyclonal | |
| Unconjugated | |
| PELO | |
| CGI 17, PELO, pelota homolog (Drosophila), PRO1770, Protein pelota homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 105083, 294754, 53918 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5XIP1, Q80X73, Q9BRX2 | |
| PELO | |
| PELO Fusion Protein Ag0873 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering