missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF R beta Antibody (APB5) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 3 publications
361.00€ - 512.00€
Specifications
| Antigen | PDGF R beta |
|---|---|
| Clone | APB5 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry < 1.0 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18296893
|
Novus Biologicals
NBP1-43349-0.05MG |
0.05 mg |
361.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18781244
|
Novus Biologicals
NBP1-43349-0.1mg |
0.1 mg |
512.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Protein Array.Specifications
| PDGF R beta | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Mouse | |
| P05622 | |
| 5159 | |
| The immunogen for this antibody was PDGF receptor beta. | |
| Primary | |
| Store at 4C. Do not freeze. |
| APB5 | |
| Flow Cytometry < 1.0 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Rat | |
| Protein Kinase | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
| PDGFRB | |
| IgG2a κ | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title