missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF R beta Antibody (APB5) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-43349-0.1mg
This item is not returnable.
View return policy
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Protein Array.
Specifications
| PDGF R beta | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry < 1.0 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| P05622 | |
| PDGFRB | |
| The immunogen for this antibody was PDGF receptor beta. | |
| 0.1 mg | |
| Protein Kinase | |
| 5159 | |
| Store at 4C. Do not freeze. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| APB5 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction