missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Antibody (7A11B1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
218.00€
Specifications
| Antigen | PD-1 |
|---|---|
| Clone | 7A11B1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL |
| Applications | Western Blot |
Description
PD-1 Monoclonal specifically detects PD-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| PD-1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| 5133 | |
| A ∼150 amino acid recombinant protein from near the amino terminus of mouse PD-1. aa 22 - 170 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 7A11B1 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Monoclonal | |
| Mouse | |
| Apoptosis, Stem Cell Markers | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| PDCD1 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title