missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Antibody (7A11B1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-75518-0.025mg
This item is not returnable.
View return policy
Description
PD-1 Monoclonal specifically detects PD-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| PD-1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| PDCD1 | |
| A ∼150 amino acid recombinant protein from near the amino terminus of mouse PD-1. aa 22 - 170 | |
| 0.025 mg | |
| Apoptosis, Stem Cell Markers | |
| 5133 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| 7A11B1 | |
| Unconjugated | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction