missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
222.00€ - 526.00€
Specifications
| Antigen | PARP7 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677731
|
Novus Biologicals
NBP2-93275-0.02ml |
0.02 mL |
222.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611461
|
Novus Biologicals
NBP2-93275-0.1ml |
0.1 mL |
526.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PARP7 Polyclonal antibody specifically detects PARP7 in Human, Mouse samples. It is validated for Western BlotSpecifications
| PARP7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS (pH 7.3), 50% glycerol | |
| 25976 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DDF1, DKFZP434J214, DKFZp686N0351, DKFZp686P1838, EC 2.4.2.30, FLJ40466, PARP-1, PARP-7, PARP7DKFZp434J214, pART14, Poly [ADP-ribose] polymerase 7, RM1, TCDD-inducible poly [ADP-ribose] polymerase, TCDD-inducible poly(ADP-ribose) polymerase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 441-657 of human PARP7 (NP_056323.2). DGVTSANFYPETWVYMHPSQDFIQVPVSAEDKSYRIIYNLFHKTVPEFKYRILQILRVQNQFLWEKYKRKKEYMNRKMFGRDRIINERHLFHGTSQDVVDGICKHNFDPRVCGKHATMFGQGSYFAKKASYSHNFSKKSSKGVHFMFLAKVLTGRYTMGSHGMRRPPPVNPGSVTSDLYDSCVDNFFEPQIFVIFNDDQSYPYFVIQYEEVSNTVSI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title