missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93275-0.02ml
This item is not returnable.
View return policy
Description
PARP7 Polyclonal antibody specifically detects PARP7 in Human, Mouse samples. It is validated for Western Blot
Specifications
| PARP7 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DDF1, DKFZP434J214, DKFZp686N0351, DKFZp686P1838, EC 2.4.2.30, FLJ40466, PARP-1, PARP-7, PARP7DKFZp434J214, pART14, Poly [ADP-ribose] polymerase 7, RM1, TCDD-inducible poly [ADP-ribose] polymerase, TCDD-inducible poly(ADP-ribose) polymerase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 441-657 of human PARP7 (NP_056323.2). DGVTSANFYPETWVYMHPSQDFIQVPVSAEDKSYRIIYNLFHKTVPEFKYRILQILRVQNQFLWEKYKRKKEYMNRKMFGRDRIINERHLFHGTSQDVVDGICKHNFDPRVCGKHATMFGQGSYFAKKASYSHNFSKKSSKGVHFMFLAKVLTGRYTMGSHGMRRPPPVNPGSVTSDLYDSCVDNFFEPQIFVIFNDDQSYPYFVIQYEEVSNTVSI | |
| 0.02 mL | |
| Apoptosis | |
| 25976 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction