missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARL Antibody (8C4B2), Novus Biologicals™
Mouse Monoclonal Antibody
168.00€
Specifications
| Antigen | PARL |
|---|---|
| Clone | 8C4B2 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
PARL Monoclonal specifically detects PARL in Human samples. It is validated for Western Blot, ELISA.Specifications
| PARL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 55486 | |
| Purified recombinant fragment of PARL (aa112-167) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 8C4B2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, Apoptosis, Mitochondrial Fusion Proteins, Neuroscience | |
| EC 3.4.21.105, mitochondrial, presenilin associated, rhomboid-like, PRO2207, rhomboid 7 homolog 1 | |
| PARL | |
| IgG2b | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title