missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARL Antibody (8C4B2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51546-0.025ml
This item is not returnable.
View return policy
Description
PARL Monoclonal specifically detects PARL in Human samples. It is validated for Western Blot, ELISA.
Specifications
| PARL | |
| Monoclonal | |
| Unconjugated | |
| EC 3.4.21.105, mitochondrial, presenilin associated, rhomboid-like, PRO2207, rhomboid 7 homolog 1 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| Western Blot | |
| 8C4B2 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| PARL | |
| Purified recombinant fragment of PARL (aa112-167) expressed in E. Coli. | |
| 0.025 mL | |
| Alzheimers Research, Apoptosis, Mitochondrial Fusion Proteins, Neuroscience | |
| 55486 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction