missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parkin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00€ - 507.00€
Specifications
| Antigen | Parkin |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645027
|
Novus Biologicals
NBP2-41287-0.025mg |
0.025 mg |
178.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18141499
|
Novus Biologicals
NBP2-41287 |
0.1 mg |
507.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Parkin Polyclonal specifically detects Parkin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| Parkin | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| E3 ubiquitin ligase, E3 ubiquitin-protein ligase parkin, EC 6.3.2.-, LPRS2, parkin, parkin 2, Parkinson disease protein 2, Parkinson juvenile disease protein 2, parkinson protein 2, E3 ubiquitin protein ligase (parkin), PDJjuvenile) 2, parkin | |
| PRKN | |
| IgG | |
| Affinity Purified | |
| 55 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| PBS with 0.02% Sodium Azide | |
| 5071 | |
| Antibody was raised against a 17 amino acid peptide near the amino terminus of human PARK2. The immunogen is located within amino acids 110 - 160 of PARK2. Amino Acid Squence: HTDSRKDSPPAGSPAG | |
| Primary | |
| PARK2 antibody is human specific. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title