missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parkin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41287-0.025mg
This item is not returnable.
View return policy
Description
Parkin Polyclonal specifically detects Parkin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Parkin | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| PRKN | |
| Antibody was raised against a 17 amino acid peptide near the amino terminus of human PARK2. The immunogen is located within amino acids 110 - 160 of PARK2. Amino Acid Squence: HTDSRKDSPPAGSPAG | |
| Affinity Purified | |
| RUO | |
| Primary | |
| PARK2 antibody is human specific. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| E3 ubiquitin ligase, E3 ubiquitin-protein ligase parkin, EC 6.3.2.-, LPRS2, parkin, parkin 2, Parkinson disease protein 2, Parkinson juvenile disease protein 2, parkinson protein 2, E3 ubiquitin protein ligase (parkin), PDJjuvenile) 2, parkin | |
| Rabbit | |
| 55 kDa | |
| 0.025 mg | |
| Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 5071 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction