missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PABPC4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38218-20ul
This item is not returnable.
View return policy
Description
PABPC4 Polyclonal antibody specifically detects PABPC4 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| PABPC4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| APP1FLJ43938, APP-1PABP-4, Inducible poly(A)-binding protein, IPABP, iPABPpoly(A)-binding protein, cytoplasmic 4 (inducible form), PABP4Activated-platelet protein 1, poly(A) binding protein, cytoplasmic 4 (inducible form), Poly(A)-binding protein 4, polyadenylate-binding protein 4 | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human PABPC4 (NP_003810.1).,, Sequence:, VTEMNGRIVGSKPLYVALAQRKEERKAHLTNQYMQRVAGMRALPANAILNQFQPAAGGYFVPAVPQAQGRPPYYTPNQLAQMRPNPRWQQGGRPQGFQGMP | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8761 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction