missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p23/PTGES3 Antibody (JJ6), Novus Biologicals™
Mouse Monoclonal Antibody
169.00€
Specifications
| Antigen | p23/PTGES3 |
|---|---|
| Clone | JJ6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18545432
|
Novus Biologicals
NB110-96879-0.025mg |
0.025 mg | |||||||
Description
p23/PTGES3 Monoclonal specifically detects p23/PTGES3 in Human, Mouse, Chicken, Guinea Pig, Rabbit, Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Microarray.Specifications
| p23/PTGES3 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones | |
| Q15185, NP_006592.3 | |
| 10728 | |
| Recombinant human full length p23 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| JJ6 | |
| Western Blot 1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cPGESp23, Cytosolic prostaglandin E2 synthase, Hsp90 co-chaperone, P23cytosolic prostaglandin E synthase, Progesterone receptor complex p23, prostaglandin E synthase 3, prostaglandin E synthase 3 (cytosolic), TEBPEC 5.3.99.3, Telomerase-binding protein p23, unactive progesterone receptor, 23 kD | |
| PTGES3 | |
| IgG1 | |
| Protein G purified | |
| Detects approx 23kDa. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title