missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p23/PTGES3 Antibody (JJ6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-96879-0.025mg
This item is not returnable.
View return policy
Description
p23/PTGES3 Monoclonal specifically detects p23/PTGES3 in Human, Mouse, Chicken, Guinea Pig, Rabbit, Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Microarray.
Specifications
| p23/PTGES3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500 | |
| cPGESp23, Cytosolic prostaglandin E2 synthase, Hsp90 co-chaperone, P23cytosolic prostaglandin E synthase, Progesterone receptor complex p23, prostaglandin E synthase 3, prostaglandin E synthase 3 (cytosolic), TEBPEC 5.3.99.3, Telomerase-binding protein p23, unactive progesterone receptor, 23 kD | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects approx 23kDa. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| JJ6 | |
| Unconjugated | |
| Q15185, NP_006592.3 | |
| PTGES3 | |
| Recombinant human full length p23 protein | |
| 0.025 mg | |
| Membrane Trafficking and Chaperones | |
| 10728 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction