Learn More
ORP1 Antibody (S02-2E2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19727-100ul
This item is not returnable.
View return policy
Description
ORP1 Monoclonal antibody specifically detects ORP1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ORP1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S02-2E2 | |
| Unconjugated | |
| FLJ10217, ORP1oxysterol-binding protein-related protein 1, ORP-1oxysterol-binding protein-related protein 1 variant 1, OSBP8, OSBPL1, OSBPL1B, OSBP-related protein 1, oxysterol binding protein-like 1A, oxysterol binding protein-like 1B, oxysterol-binding protein-related protein 1 variant 2 | |
| A synthetic peptide of human ORP1 (Uniprot # Q9BXW6) | |
| 100 μg | |
| Lipid and Metabolism | |
| 114876 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |