missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC1 Antibody (ORC1 7F6/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
214.00€
Specifications
| Antigen | ORC1 |
|---|---|
| Clone | ORC1 7F6/1 |
| Concentration | 0.76 mg/ml |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
ORC1 Monoclonal specifically detects ORC1 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunoprecipitation.Specifications
| ORC1 | |
| 0.76 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 4998 | |
| Bacterially expressed Human ORC1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| ORC1 7F6/1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| HS, HSORC1, ORC1Lorigin recognition complex subunit 1, origin recognition complex, subunit 1, origin recognition complex, subunit 1 (yeast homolog)-like, origin recognition complex, subunit 1 homolog, origin recognition complex, subunit 1 homolog (S. cerevisiae), origin recognition complex, subunit 1-like, origin recognition complex, subunit 1-like (yeast), PARC1origin recognition complex 1, Replication control protein 1 | |
| ORC1 | |
| IgG1 | |
| Protein A purified | |
| ORC1 (7F6/1). Cross-reacts with Human. Not yet tested in other species. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title