missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC1 Antibody (ORC1 7F6/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-121-0.025mg
This item is not returnable.
View return policy
Description
ORC1 Monoclonal specifically detects ORC1 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunoprecipitation.
Specifications
| ORC1 | |
| Monoclonal | |
| 0.76 mg/ml | |
| HS, HSORC1, ORC1Lorigin recognition complex subunit 1, origin recognition complex, subunit 1, origin recognition complex, subunit 1 (yeast homolog)-like, origin recognition complex, subunit 1 homolog, origin recognition complex, subunit 1 homolog (S. cerevisiae), origin recognition complex, subunit 1-like, origin recognition complex, subunit 1-like (yeast), PARC1origin recognition complex 1, Replication control protein 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| ORC1 (7F6/1). Cross-reacts with Human. Not yet tested in other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| ORC1 7F6/1 | |
| Unconjugated | |
| ORC1 | |
| Bacterially expressed Human ORC1 | |
| 0.025 mg | |
| Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4998 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction