missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orai1 Antibody (6D11A11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
209.00€
Specifications
| Antigen | Orai1 |
|---|---|
| Clone | 6D11A11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL |
| Applications | Western Blot |
Description
Orai1 Monoclonal specifically detects Orai1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| Orai1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| CRACM1ORAT1, FLJ14466, ORAI calcium release-activated calcium modulator 1, Protein orai-1, TMEM142Acalcium release-activated calcium modulator 1, Transmembrane protein 142Acalcium release-activated calcium channel protein 1 | |
| ORAI1 | |
| IgG2b | |
| Affinity Purified | |
| ORAI1 antibody is predicted to have no cross-reactivity to ORAI2 or ORAI3. |
| 6D11A11 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| 84876 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human ORAI1. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title