missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orai1 Antibody (6D11A11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-75523-0.025mg
This item is not returnable.
View return policy
Description
Orai1 Monoclonal specifically detects Orai1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Orai1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| ORAI1 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human ORAI1. | |
| Affinity Purified | |
| RUO | |
| 84876 | |
| Human | |
| IgG2b |
| Western Blot | |
| 6D11A11 | |
| Unconjugated | |
| CRACM1ORAT1, FLJ14466, ORAI calcium release-activated calcium modulator 1, Protein orai-1, TMEM142Acalcium release-activated calcium modulator 1, Transmembrane protein 142Acalcium release-activated calcium channel protein 1 | |
| Mouse | |
| 50 kDa | |
| 0.025 mg | |
| Primary | |
| ORAI1 antibody is predicted to have no cross-reactivity to ORAI2 or ORAI3. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction