missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Optineurin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-26700
This item is not returnable.
View return policy
Description
Optineurin Polyclonal antibody specifically detects Optineurin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Optineurin | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| FIP2FIP-2, GLC1Eglaucoma 1, open angle, E (adult-onset), HIP7ALS12, Huntingtin interacting protein L, Huntingtin yeast partner L, Huntingtin-interacting protein 7, Huntingtin-interacting protein L, HYPLHIP-7, NEMO-related protein, NRPE3-14.7K-interacting protein, Optic neuropathy-inducing protein, optineurin, TFIIIA-INTP, Transcription factor IIIA-interacting protein, transcrption factor IIIA-interacting protein, tumor necrosis factor alpha-inducible cellular protein containing leucinezipper domains | |
| Partial recombinant protein made to an internal portion of human Optineurin between amino acids 150-350 [UniProt Q96CV9]. | |
| 0.1 mg | |
| Immunology, Innate Immunity, Vision | |
| 10133 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| 1.0 mg/mL | |
| Western Blot 1.0 μg/mL, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1.0 μg/mL, Immunohistochemistry-Paraffin 1:200 | |
| Q96CV9 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction