missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Optineurin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
171.00€ - 451.00€
Specifications
| Antigen | Optineurin |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1.0 μg/mL, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1.0 μg/mL, Immunohistochemistry-Paraffin 1:200 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043881
|
Novus Biologicals™
NBP3-26700 |
0.1 mg |
451.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043283
|
Novus Biologicals™
NBP3-26700SS |
0.025 mg |
171.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Optineurin Polyclonal antibody specifically detects Optineurin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ ImmunofluorescenceSpecifications
| Optineurin | |
| Western Blot 1.0 μg/mL, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1.0 μg/mL, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Q96CV9 | |
| 10133 | |
| IgG | |
| Immunogen affinity purified |
| 1.0 mg/mL | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity, Vision | |
| PBS | |
| FIP2FIP-2, GLC1Eglaucoma 1, open angle, E (adult-onset), HIP7ALS12, Huntingtin interacting protein L, Huntingtin yeast partner L, Huntingtin-interacting protein 7, Huntingtin-interacting protein L, HYPLHIP-7, NEMO-related protein, NRPE3-14.7K-interacting protein, Optic neuropathy-inducing protein, optineurin, TFIIIA-INTP, Transcription factor IIIA-interacting protein, transcrption factor IIIA-interacting protein, tumor necrosis factor alpha-inducible cellular protein containing leucinezipper domains | |
| Partial recombinant protein made to an internal portion of human Optineurin between amino acids 150-350 [UniProt Q96CV9]. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title