Learn More
Oil and grease cncentrate, QC sample, 23mL, SPEX CertiPrep™
Product Code. 11984974 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
23 mL
Unit Size:
23mL
Product Code. 11984974
Brand: SPEX Certiprep 4122