missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCIL/CLEC2d Antibody (166C1309), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56553SS
This item is not returnable.
View return policy
Description
OCIL/CLEC2d Monoclonal specifically detects OCIL/CLEC2d in Mouse samples. It is validated for Western Blot.
Specifications
| OCIL/CLEC2d | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot | |
| CLAXLLT-1, C-type lectin domain family 2 member D, C-type lectin domain family 2, member D, C-type lectin related f, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT1Osteoclast inhibitory lectin, OCILmember D | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| 166C1309 | |
| Unconjugated | |
| AAK50881 | |
| CLEC2D | |
| This antibody was developed by immunizing mice with a synthetic peptide containing amino acids 2-18 (CVTKASLPMLSPTGSPQ) of mouse OCIL, GenBank no. gb AAL37198.1. | |
| 0.025 mg | |
| Immunology | |
| 29121 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction