missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-GlcNAcase/OGA/MGEA5 Antibody (8D12), Novus Biologicals™
Rabbit Monoclonal Antibody
355.00€ - 568.00€
Specifications
| Antigen | O-GlcNAcase/OGA/MGEA5 |
|---|---|
| Clone | 8D12 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30038959
|
Novus Biologicals
NBP3-26142-50ul |
50 μL |
355.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30031951
|
Novus Biologicals
NBP3-26142-100ul |
100 μL |
568.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
O-GlcNAcase/OGA/MGEA5 Monoclonal antibody specifically detects O-GlcNAcase/OGA/MGEA5 in Human samples. It is validated for Western Blot, ELISASpecifications
| O-GlcNAcase/OGA/MGEA5 | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 10724 | |
| IgG | |
| Affinity purified |
| 8D12 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| bifunctional protein NCOAT, FLJ11229, HEXC, hyaluronidase in meningioma, KIAA0679, MEA5FLJ23355, meningioma expressed antigen 5 (hyaluronidase), Meningioma-expressed antigen 5, NCOAT, Nuclear cytoplasmic O-GlcNAcase and acetyltransferase, OGA, O-GlcNAcase | |
| A synthesized peptide derived from Human O-GlcNAcase/OGA/MGEA5 [UniProt O60502] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title