missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-GlcNAcase/OGA/MGEA5 Antibody (8D12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26142-100ul
This item is not returnable.
View return policy
Description
O-GlcNAcase/OGA/MGEA5 Monoclonal antibody specifically detects O-GlcNAcase/OGA/MGEA5 in Human samples. It is validated for Western Blot, ELISA
Specifications
| O-GlcNAcase/OGA/MGEA5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10724 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| 8D12 | |
| Western Blot 1:500-1:5000, ELISA | |
| bifunctional protein NCOAT, FLJ11229, HEXC, hyaluronidase in meningioma, KIAA0679, MEA5FLJ23355, meningioma expressed antigen 5 (hyaluronidase), Meningioma-expressed antigen 5, NCOAT, Nuclear cytoplasmic O-GlcNAcase and acetyltransferase, OGA, O-GlcNAcase | |
| A synthesized peptide derived from Human O-GlcNAcase/OGA/MGEA5 [UniProt O60502] | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction