missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT18 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94736-0.02ml
This item is not returnable.
View return policy
Description
NUDT18 Polyclonal antibody specifically detects NUDT18 in Human samples. It is validated for Western Blot
Specifications
| NUDT18 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 3.6.1, EC 3.6.1.-, FLJ22494, nucleoside diphosphate-linked moiety X motif 18, nudix (nucleoside diphosphate linked moiety X)-type motif 18, Nudix motif 18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-323 of human NUDT18 (NP_079091.3). MASEGLAGALASVLAGQGSSVHSCDSAPAGEPPAPVRLRKNVCYVVLAVFLSEQDEVLLIQEAKRECRGSWYLPAGRMEPGETIVEALQREVKEEAGLHCEPETLLSVEERGPSWVRFVFLARPTGGILKTSKEADAESLQAAWYPRTSLPTPLRAHDILHLVELAAQYRQQARHPLILPQELPCDLVCQRLVATFTSAQTVWVLVGTVGMPHLPVTACGLDPMEQRGGMKMAVLRLLQECLTLHHLVVEIKGLLGLQHLGRDHSDGICLNVLVTVAFRSPGIQDEPPKVRGENFSWWKVMEEDLQSQLLQRLQGSSVVPVNR | |
| 0.02 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 79873 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction