missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT18 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 482.00€
Specifications
| Antigen | NUDT18 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643742
|
Novus Biologicals
NBP2-94736-0.02ml |
0.02 mL |
190.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679531
|
Novus Biologicals
NBP2-94736-0.1ml |
0.1 mL |
482.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NUDT18 Polyclonal antibody specifically detects NUDT18 in Human samples. It is validated for Western BlotSpecifications
| NUDT18 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 79873 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.6.1, EC 3.6.1.-, FLJ22494, nucleoside diphosphate-linked moiety X motif 18, nudix (nucleoside diphosphate linked moiety X)-type motif 18, Nudix motif 18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-323 of human NUDT18 (NP_079091.3). MASEGLAGALASVLAGQGSSVHSCDSAPAGEPPAPVRLRKNVCYVVLAVFLSEQDEVLLIQEAKRECRGSWYLPAGRMEPGETIVEALQREVKEEAGLHCEPETLLSVEERGPSWVRFVFLARPTGGILKTSKEADAESLQAAWYPRTSLPTPLRAHDILHLVELAAQYRQQARHPLILPQELPCDLVCQRLVATFTSAQTVWVLVGTVGMPHLPVTACGLDPMEQRGGMKMAVLRLLQECLTLHHLVVEIKGLLGLQHLGRDHSDGICLNVLVTVAFRSPGIQDEPPKVRGENFSWWKVMEEDLQSQLLQRLQGSSVVPVNR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title