missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoType Seed PCR
Kit designed for rapid genotyping from seed samples.
27.60€ - 386.00€
Specifications
Content And Storage | Lysis Buffer P; Proteinase K; NucleoType HotStart PCR Master Mix (2x) containing polymerase, dNTPs, buffer, enhancer, loading dye and stabilizer. Store at 4°C or -20°C |
---|---|
Format | Kit |
Reaction Speed | 30 to 90 min. |
Kit Type | Simple sample preparation suitable for hot start PCR |
For Use With (Application) | Genotyping |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
16462185
|
Macherey-Nagel™
743203.25 |
25 |
27.60€
25 reactions |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16447595
|
Macherey-Nagel™
743203.100 |
100 |
87.00€
100 reactions |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16472185
|
Macherey-Nagel™
743203.500 |
250 |
386.00€
500 reactions |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
Description
- Preparation within 5 minutes
- No need for DNA purification
- Loading dye and Proteinase K included
- Easy genoytyping of whole and ground seeds
Specifications
Lysis Buffer P; Proteinase K; NucleoType HotStart PCR Master Mix (2x) containing polymerase, dNTPs, buffer, enhancer, loading dye and stabilizer. Store at 4°C or -20°C | |
30 to 90 min. | |
Genotyping | |
Amplicon size up to 2000 bp |
Kit | |
Simple sample preparation suitable for hot start PCR | |
Hard plant material like e.g., seeds from soybeen, wheat, corn, rice, as well as from moss, fern leaf, and fir needle |