Learn More
Macherey-Nagel™ NucleoType Seed PCR
Kit designed for rapid genotyping from seed samples.
Brand: Macherey-Nagel™ 743203.100
Additional Details : Weight : 0.07000kg
Description
- Preparation within 5 minutes
- No need for DNA purification
- Loading dye and Proteinase K included
- Easy genoytyping of whole and ground seeds
Specifications
Lysis Buffer P; Proteinase K; NucleoType HotStart PCR Master Mix (2x) containing polymerase, dNTPs, buffer, enhancer, loading dye and stabilizer. Store at 4°C or -20°C | |
30 to 90 min. | |
Genotyping | |
Hard plant material like e.g., seeds from soybeen, wheat, corn, rice, as well as from moss, fern leaf, and fir needle |
Kit | |
Simple sample preparation suitable for hot start PCR | |
100 | |
Amplicon size up to 2000 bp |