Learn More
Macherey-Nagel™ NucleoType Seed PCR
Kit designed for rapid genotyping from seed samples.
Brand:  Macherey-Nagel™ 743203.25
Additional Details : Weight : 0.01500kg
 View more versions of this product
Product Code. 16462185
  • 27.60€ / 25 reactions
Estimated Shipment: 28-06-2024
Description
  • Preparation within 5 minutes
  • No need for DNA purification
  • Loading dye and Proteinase K included
  • Easy genoytyping of whole and ground seeds
Specifications
Specifications
Lysis Buffer P; Proteinase K; NucleoType HotStart PCR Master Mix (2x) containing polymerase, dNTPs, buffer, enhancer, loading dye and stabilizer. Store at 4°C or -20°C
30 to 90 min.
Genotyping
Hard plant material like e.g., seeds from soybeen, wheat, corn, rice, as well as from moss, fern leaf, and fir needle
Kit
Simple sample preparation suitable for hot start PCR
25
Amplicon size up to 2000 bp