missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V delta 2 Antibody (15D), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22508-50ug
This item is not returnable.
View return policy
Description
TCR V delta 2 Monoclonal specifically detects TCR V delta 2 in Human, Drosophila, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| TCR V delta 2 | |
| Monoclonal | |
| 0.15 mg/mL | |
| Western Blot 1:1 - 1:100, Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional | |
| TRD | |
| Human TCR V delta 2 | |
| 50 μg | |
| Primary | |
| This antibody reacts with Human. Drosophila reactivity reported in scientific literature (PMID: 18802083). Rhesus Monkey reactivity reported in scientific literature (PMID: 20660793). | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay | |
| 15D | |
| Unconjugated | |
| T cell receptor delta locus, T-cell antigen receptor, delta polypeptide, T-cell receptor, delta (V,D,J,C), TCRD, TCRD, TRD@, TCRDV1, TRD@ | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6964 | |
| Human, Drosophila, Primate | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction